Order Entry
Puerto Rico
ContactUsLinkComponent
InVivoMAb Anti-osteopontin (SPP1) Mouse Monoclonal Antibody [clone: MPIIIB10]
InVivoMAb Anti-osteopontin (SPP1) Mouse Monoclonal Antibody [clone: MPIIIB10]
Supplier:  Bio X Cell
New Product
Certificates
InVivoMAb Anti-osteopontin (SPP1) Mouse Monoclonal Antibody [clone: MPIIIB10]
Supplier:  Bio X Cell
New Product

Specifications

  • Catalog No:
  • 77579-368
  • 77579-366
  • 77579-364
  • 77579-362
  • 77579-360
  • Antigen Symbol:
  • osteopontin (SPP1)
  • osteopontin (SPP1)
  • osteopontin (SPP1)
  • osteopontin (SPP1)
  • osteopontin (SPP1)
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • MPIIIB10
  • MPIIIB10
  • MPIIIB10
  • MPIIIB10
  • MPIIIB10
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Tested Applications:
  • in vivo OPN neutralization, in vitro OPN neutralization, Immunohistochemistry (paraffin)
  • in vivo OPN neutralization, in vitro OPN neutralization, Immunohistochemistry (paraffin)
  • in vivo OPN neutralization, in vitro OPN neutralization, Immunohistochemistry (paraffin)
  • in vivo OPN neutralization, in vitro OPN neutralization, Immunohistochemistry (paraffin)
  • in vivo OPN neutralization, in vitro OPN neutralization, Immunohistochemistry (paraffin)

Specifications