Metal Detectable Long Handled Spoon
Catalog # 77578-214
Supplier: SAMPLING SYSTEMS LTD
New Product
Specifications
- Capacity:20 ml
- Color:Blue
- Description:Long handled spoon straight, metal detectable PP
- Cat. No.:77578-214
Specifications
About this item
Powder sampling is easy with our metal detectable spoons. They have a very long handle which means they are easy to see if left in the powder. They are made from a special robust, chemically resistant grade of PP.
- Easy to use
- Made from high quality PP, conforms to the latest FDA and EU food requirements
- 20 ml volume is a standard size
They are moulded in a Class 100000 cleanroom to ensure the cleanliness of the product. The robust design means they can be used for years and years or used as a single use product straight from the bag.
Simply unwrap and use. They are ideal for use with liquids, creams and pastes. The blue colour means they can easily seen if left in the material in line with HACCP guidelines.