ContactUsLinkComponent
Screw Cap for Urine Collection Cups
Catalog # 77589-086
Supplier:  SARSTEDT INC
New Product
undefined
Screw Cap for Urine Collection Cups
Catalog # 77589-086
Supplier:  SARSTEDT INC
Supplier Number:  76.564
New Product
Specifications
  • Color:
    Natural
  • Description:
    Screw cap for 100 ml urine collection cup, 14Ø×62H mm
  • Material:
    HDPE
  • Cat. No.:
    77589-086
  • Packaging:
    BAGA
Specifications
About this item
To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: HDPE
  • Compatible with screw cap for 100 ml urine cup (77589-064)
  • Odor-tight screw cap
  • For secure transport and storage of urine samples
Small volume urine containers for consistent and reliable samples.