Order Entry
Puerto Rico
ContactUsLinkComponent
Blood Gas Capillaries
Blood Gas Capillaries
Catalog # 77587-416
Supplier:  SARSTEDT INC
New Product
undefined
Blood Gas Capillaries
Catalog # 77587-416
Supplier:  SARSTEDT INC
Supplier Number:  19.930.175
New Product

Specifications

  • Ø:
    2.3 mm
  • Volume:
    175 µl
  • Description:
    Blood gas capillaries with calcium-balanced lithium heparin preparation
  • Length:
    100 mm
  • Material:
    PET
  • Cat. No.:
    77587-416
  • Packaging:
    Container

Specifications

About this item

For small sample volumes and optimal instrument compatibility, plastic blood gas capillaries are available in a variety of sizes and volumes.

  • For capillary blood gas analysis with range of volumes
  • Preparation: Calcium-balanced lithium heparin in droplet form as a spray dose; 80 IU heparin/ml blood
  • Calcium (Ca²⁺) balanced heparin is also suitable for determining electrolytes
  • Low gas permeable PET is flexible and robust

Blood gas capillaries provide reliable measurement results with a special calcium-balanced heparin coating. Break-resistant PET plastic ensures maximum safety during collection. The special surface treatment enables quick filling of the capillary, making sample collection easier and reducing the risk of coagulation.