CSF Tubes
Proven material properties specifically for CSF samples.
- Base shape: Round base
- Tight-closing screw cap for transport and storage
- Common tube dimensions suitable for automated processing
- With graduations to check the filling level
Clear and sterile collection tubes are particularly necessary for CSF diagnostics. The use of appropriate materials such as polypropylene helps to ensure standardized collection of CSF for pre-analytics.
Proven material properties specifically for CSF samples.
- Base shape: Round base
- Tight-closing screw cap for transport and storage
- Common tube dimensions suitable for automated processing
- With graduations to check the filling level
Clear and sterile collection tubes are particularly necessary for CSF diagnostics. The use of appropriate materials such as polypropylene helps to ensure standardized collection of CSF for pre-analytics.
Product Family Options
- DescriptionMaterialØ×LVolumeCap colorCap materialGraduationsPackagingSterility
- CSF tubes with non-writable, transparent/green labelPP13×75 mm5 mlNaturalHDPEWith graduationsBlisterSterileCase of 500$296.69: 63.504.027
- CSF tubes with non-writable, green labelPP15.3×92 mm10 mlNaturalHDPEWith graduationsBlisterSterileCase of 500$245.83: 62.610.018