Prepared Micro Sample Tubes with Push Caps
Supplier: SARSTEDT INC
New Product
Certificates
About this item
SARSTEDT prepared micro tubes are available with push cap closure or screw cap with integral O-ring.
- Ø: 10.8 mm
- Length without/with cap: 40/44 mm
- Base: Skirted conical base
- Ideal collection vessel for small volumes of blood
- Attached push cap suitable for one-handed collection technique
- Transparent label with graduations ideal for checking fill level; available with paper label (*.015) on request
These micro volume tubes are prepared with standard additives for versatile low-volume requirements.