Microvette® 300 Capillary Blood Collection Tubes with Collection Rim
New Product
Capillary blood collection facilitates diagnostics with a small volume of blood. SARSTEDT offers innovative solutions for all patient groups.
- Closure: HDPE cap
- Outer container with skirted base
- Smooth cap - special lid design reduces the aerosol effect
- Graduations to check fill level
- Conical inner tube: Supernatant is higher after centrifugation for easier collection of plasma
- Collection via collection rim
The capillary blood collection systems are among the most versatile on the market. Microvette® with nominal volumes between 100 and 500 µl, conical or cylindrical inner shapes, as well as choice of collection technique end-to-end or with a collection rim.
Shape of the inner tube: Conical
Base shape: Flat base
Frequently Bought Together