RecombiMAb Human IgG4 (S228P/R409K) Isotype control Anti-Hen Egg Lysozyme
New Product
Type: Control
Antigen: IgG4
Clonality: Recombinant
Clone: N/A-CP183
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Hen
- Catalog No:
- 77580-488
- 77580-486
- 77580-484
- 77580-482
- 77580-480
- Antigen Symbol:
- IgG4
- IgG4
- IgG4
- IgG4
- IgG4
- Antibody Type:
- Control
- Control
- Control
- Control
- Control
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- N/A-CP183
- N/A-CP183
- N/A-CP183
- N/A-CP183
- N/A-CP183