RecombiMAb Mouse IgG2a Isotype control Unknown Specificity
Supplier: Bio X Cell
About this item
Type: Control
Antigen: IgG1 ?
Clonality: Recombinant
Clone: MOPC-21-CP160
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Control
Antigen: IgG1 ?
Clonality: Recombinant
Clone: MOPC-21-CP160
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Specifications for Product Family
Specifications
- Catalog No:
- 77580-352
- 77580-358
- 77580-354
- 77580-356
- 77580-350
- Antigen Symbol:
- IgG1 ?
- IgG1 ?
- IgG1 ?
- IgG1 ?
- IgG1 ?
- Antibody Type:
- Control
- Control
- Control
- Control
- Control
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- MOPC-21-CP160
- MOPC-21-CP160
- MOPC-21-CP160
- MOPC-21-CP160
- MOPC-21-CP160
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
- Yes
- Tested Applications:
- in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
- in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
- in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
- in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
- in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: CP160-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77580-352Antigen SymbolIgG1 ?Antibody TypeControlClonalityRecombinantCloneMOPC-21-CP160Western BlotYesFlow CytometryYesTested Applicationsin vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody - Supplier Number: CP160-5MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77580-358Antigen SymbolIgG1 ?Antibody TypeControlClonalityRecombinantCloneMOPC-21-CP160Western BlotYesFlow CytometryYesTested Applicationsin vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody - Supplier Number: CP160-25MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77580-354Antigen SymbolIgG1 ?Antibody TypeControlClonalityRecombinantCloneMOPC-21-CP160Western BlotYesFlow CytometryYesTested Applicationsin vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody - Supplier Number: CP160-50MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77580-356Antigen SymbolIgG1 ?Antibody TypeControlClonalityRecombinantCloneMOPC-21-CP160Western BlotYesFlow CytometryYesTested Applicationsin vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody - Supplier Number: CP160-100MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77580-350Antigen SymbolIgG1 ?Antibody TypeControlClonalityRecombinantCloneMOPC-21-CP160Western BlotYesFlow CytometryYesTested Applicationsin vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



