InVivoMAb Anti-InVivoMAb anti-mouse/human osteopontin (SPP1) Mouse Monoclonal Antibody [clone: 100D3]
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Primary
Antigen: InVivoMAb anti-mouse/human osteopontin (SPP1)
Clonality: Monoclonal
Clone: 100D3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse;Human
Specifications
- Catalog No:
- 77579-268
- 77579-266
- 77579-264
- 77579-262
- 77579-260
- Antigen Symbol:
- InVivoMAb anti-mouse/human osteopontin (SPP1)
- InVivoMAb anti-mouse/human osteopontin (SPP1)
- InVivoMAb anti-mouse/human osteopontin (SPP1)
- InVivoMAb anti-mouse/human osteopontin (SPP1)
- InVivoMAb anti-mouse/human osteopontin (SPP1)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 100D3
- 100D3
- 100D3
- 100D3
- 100D3
- Reactivity:
- Human, Mouse
- Human, Mouse
- Human, Mouse
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- ELISA:
- Yes
- Yes
- Yes
- Yes
- Yes
- Tested Applications:
- in vivo OPN neutralization, in vitro OPN neutralization
- in vivo OPN neutralization, in vitro OPN neutralization
- in vivo OPN neutralization, in vitro OPN neutralization
- in vivo OPN neutralization, in vitro OPN neutralization
- in vivo OPN neutralization, in vitro OPN neutralization