InVivoMAb Anti-amyloid-beta Mouse Monoclonal Antibody [clone: MOAB-2]
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Primary
Antigen: amyloid-beta
Clonality: Monoclonal
Clone: MOAB-2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77579-226
- 77579-224
- 77579-222
- 77579-220
- 77579-228
- Antigen Symbol:
- amyloid-beta
- amyloid-beta
- amyloid-beta
- amyloid-beta
- amyloid-beta
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- MOAB-2
- MOAB-2
- MOAB-2
- MOAB-2
- MOAB-2
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Yes
- Tested Applications:
- Immunohistochemistry
- Immunohistochemistry
- Immunohistochemistry
- Immunohistochemistry
- Immunohistochemistry