NANOCOLOR® Photometric Tube Tests, Nonionic Surfactants
Catalog # 77578-530
Supplier: Macherey-Nagel
New Product
CAS Number:
Specifications
- Chemical method:Tetrabromphenolphthaleinethylester (TBE)
- Description:Nonionic surfactants 15
- Cat. No.:77578-530
- Range:0.3 - 15.0 mg/L Triton® X-100
- No. of tests:20
Specifications
About this item
A pre-dosed NANOCOLOR® tube test for photometric determination of non ionic surfactants in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.