NANOCOLOR® Photometric Tube Tests, Cationic Surfactants
Catalog # 77578-442
Supplier: Macherey-Nagel
New Product
CAS Number:
Specifications
- Chemical method:Disulfine blue
- Description:Cationic surfactants 4
- Cat. No.:77578-442
- Range:0.20 - 4.00 mg/L CTAB
- No. of tests:20
Specifications
About this item
A pre-dosed NANOCOLOR tube test for photometric determination of cationic surfactants in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in ext.Ø 16 mm tubes