Order Entry
Puerto Rico
ContactUsLinkComponent
2PACz ≥98.0% (by HPLC, titration analysis)
Irritant
2PACz ≥98.0% (by HPLC, titration analysis)
Catalog # TCC3663-500MG
Supplier:  TCI America
New Product
CAS Number:  
Irritant
2PACz ≥98.0% (by HPLC, titration analysis)
Catalog # TCC3663-500MG
Supplier:  TCI America
Supplier Number:  C3663-500MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 mg
  • Cat. No.:
    TCC3663-500MG

Specifications

Specification Test Results

  • Appearance:
    White to Blue powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :275.24 g/mol
 :GHS07
CAS Number:20999-38-6
Formula:C₁₄H₁₄NO₃P