Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:1 Vial
- Cat. No.:TCR0239-1VIAL
Specifications
About this item
The rGRFT is an Escherichia coli recombinant of carbohydrate-binding protein (Lectin) derived from Griffithsia sp. This lectin preferably binds to a [Manα(1-2)Man] glycan epitope.
The LecBeads is a lectin-immobilized agarose which is capable of capturing glycoproteins via binding to its specific epitope(s), and it is prepared by a covalent coupling method between lectin and agarose.
The Lectin-catch method using LecBeads (as shown in Application tab) is available for selective separation of glycoconjugates (glycoproteins, glycolipids, and so on). This rGRFT-LecBeads (1 ml gel) shows a binding capacity of about 6 mg or more bovine Ribonuclease B [RNase B] through its binding to a terminal [Manα(1-2)Man] glycan epitope on RNase B.
- Preparation : Recombinant GRFT is covalently coupled with agarose gel
- Quantity : 1 ml gel/2 ml (1:1, v/v slurry) suspended in PBS, pH7.5
- Protein concentration : about 10 mg/ml gel
- Binding capacity : >6 mg of glycoprotein (bovine RNase B) /ml gel
- Preservative : 0.02% [w/v] Sodium azide
- Stabilizer : None
- Store at 2-10°C. (Avoid freezing).