Order Entry
Puerto Rico
ContactUsLinkComponent
rSRL-LecBeads [for GlcNAcβ(1-2)Man, Galβ(1-3)GalNAc]
rSRL-LecBeads [for GlcNAcβ(1-2)Man, Galβ(1-3)GalNAc]
  TCR0238-1VIAL
New Product
 :  
rSRL-LecBeads [for GlcNAcβ(1-2)Man, Galβ(1-3)GalNAc]
  TCR0238-1VIAL
 :  R0238-1VIAL
New Product
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pack type:
    1 Vial
  • Cat. No.:
    TCR0238-1VIAL

 

 

The rSRL is an Escherichia coli recombinant of carbohydrate-binding protein (Lectin) derived from Sclerotium rolfsii. This lectin preferably binds to [GlcNAcβ(1-2)Man, Galβ(1-3)GalNAc] glycan epitopes by its two distinct carbohydrate-binding sites.
The LecBeads is a lectin-immobilized agarose which is capable of capturing glycoproteins via binding to its specific epitope(s), and it is prepared by a covalent coupling method between lectin and agarose.
The Lectin-catch method using LecBeads (as shown in Application tab) is available for selective separation of glycoconjugates (glycoproteins, glycolipids, and so on). This rSRL-LecBeads (1 ml gel) shows a binding capacity of about 7.5 mg or more agalacto human Transferrin [agalacto hTF] through its binding to a terminal [GlcNAcβ(1-2)Man] glycan epitope on agalacto hTF.

  • Preparation : Recombinant SRL is covalently coupled with agarose gel
  • Quantity : 1 ml gel/2 ml (1:1, v/v slurry) suspended in PBS, pH7.5
  • Protein concentration : about 15 mg/ml gel
  • Binding capacity : >7.5 mg of glycoprotein (agalacto human Transferrin) /ml gel
  • Preservative : 0.02% [w/v] Sodium azide
  • Stabilizer : None
  • Store at 2-10 °C. (Avoid freezing).