Order Entry
Puerto Rico
ContactUsLinkComponent
 
Reference Material of Column Resolution Check Mix, Certified by The Japan Petroleum Institute, TCI America
Catalog # TCS0710-1SET
Supplier:  TCI America
New Product
CAS Number:  
Reference Material of Column Resolution Check Mix, Certified by The Japan Petroleum Institute, TCI America
Catalog # TCS0710-1SET
Supplier:  TCI America
Supplier Number:  S0710-1SET
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Storage Temperature:
    Room Temperature (Recommended in a cool and dark place, <15°C)
  • Description:
    Purity (HPLC): min. 95.0 area%
    Purity(GC): min. 95.0%
  • Physical Form:
    Liquid
  • Cat. No.:
    TCS0710-1SET

Specifications

About this item

This reference material is used to confirm the peak separation. It is a mixture of the following components shown in Table below.

Methanol
2-Methylpentane
n-Butane
Isobutane
ETBE
n-Pentane
Isobutylene
3,3-Dimethyl-1-pentene
n-Hexane
Ethanol
1-Methylcyclopentene
n-Heptane
3-Methyl-1-butene
Benzene
n-Octane
cis-2-Pentene
p-Xylene
n-Nonane
trans-2-Pentene
2,3-Dimethylheptane
n-Decane
tert-Butanol
5-Methylnonane
n-Undecane
2-Methyl-2-butene
1,2-Methylethylbenzene
n-Dodecane
2,3-Dimethylbutane
1-Methylnaphthalene
n-Tridecane
MTBE
n-Propane
n-Tetradecane

Certifications: Certified by The Japan Petroleum Institute