Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:1 Vial
- Description:Protein A Agarose
- Cat. No.:TCP2461-1VIAL
Protein A is a type I membrane protein produced by several strains of Staphylococcus aureus. It has high-affinity binding sites that enable the purification of IgG obtained from various species such as humans, rabbit, mouse, and bovine. Protein A Agarose is prepared using a covalent coupling method that maintains the stability and binding characteristics of the product.
By using our resin, human IgGs can be eluted under milder conditions (such as at pH 4.0) as compared to elution by conventional methods.
Additional information is available on our website.
- Volume/Quantity : 2 ml
- Product Form : Protein A agarose is supplied pre-swollen in 20% ethanol.
- Preparation : Recombinant Protein A is covalently coupled to agarose gel.
- Preservative : 20% ethanol
Protein A agarose can be used antibody purification.
Protein A agarose should be stored in neutral pH at 2-8 °C in 20% ethanol or other bacteriostatic agent, e.g. sodium azide. The resin must not be frozen.