
2-Amino-3,7-dihydropyrrolo[2,3-d]pyrimidin-4-one 90+%
Catalog # TS450260010
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:1 g
- Cat. No.:TS450260010
Specifications
Specification Test Results
- Appearance:Grey to brown Powder
- Infrared spectrum:Conforms
- HPLC:≥90.0 %
Specification Test Results
About this item
:
:150.14 g/mol
Melting Pt:>230 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:7355-55-7
:MFCD09033259
Formula:C₆H₆N₄O