
2-Chloro-4-methyl-3-nitropyridine 98%
Catalog # TS379450050
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:5 g
- Cat. No.:TS379450050
Specifications
Specification Test Results
- Appearance:Yellow to beige Powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
About this item
:
Melting Pt:51…53 °C
Flash Pt:113 °C
:172.57 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:23056-39-5
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD00012347