Annexin V Binding Buffer
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Assay buffer for labeling with Annexin V - 50 ml.
Annexin V Binding Buffer is an isotonic buffer used in conjunction with Annexin V dyes (e.g. Catalog #100-0329) and either 7-AAD (Catalog #75001) or Propidium Iodide (Catalog #75002). This buffer contains an optimal concentration of calcium that facilitates the binding of Annexin V to phosphatidylserine on the cell surface, which is essential for the detection of early-stage cell apoptosis by flow cytometry.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
- Description:Annexin V binding buffer
- Size:50 ml