ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Thiocyanate
NANOCOLOR® Photometric Tube Tests, Thiocyanate
Catalog # 77578-580
Supplier:  Macherey-Nagel
New Product
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Thiocyanate
Catalog # 77578-580
Supplier:  Macherey-Nagel
Supplier Number:  985091
New Product
CAS Number:  
Specifications
  • Chemical method:
    Iron (III) ions
  • Description:
    Thiocyanate 50
  • Cat. No.:
    77578-580
  • Range:
    0.5 - 50.0 mg/L SCN
  • No. of tests:
    20
Specifications
About this item
A pre-dosed NANOCOLOR® tube test for photometric determination of thiocyanate in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.