Order Entry
Puerto Rico
ContactUsLinkComponent
Acetonitrile-[D₃] 100 atom % D for NMR spectroscopy
Acetonitrile-[D₃] 100 atom % D for NMR spectroscopy
Catalog # TS464170075
New Product
CAS Number:  
Acetonitrile-[D₃] 100 atom % D for NMR spectroscopy
Catalog # TS464170075
Supplier Number:  464170075
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass ampoule
  • Size:
    0.75 ml
  • Cat. No.:
    TS464170075

Specifications

About this item

 :
 :44.03 g/mol
UN:1648
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881