Order Entry
Puerto Rico
ContactUsLinkComponent
1,2-Dichlorobenzene-[D₄] 99 atom % D for NMR spectroscopy
1,2-Dichlorobenzene-[D₄] 99 atom % D for NMR spectroscopy
Catalog # TS464060010
New Product
CAS Number:  
1,2-Dichlorobenzene-[D₄] 99 atom % D for NMR spectroscopy
Catalog # TS464060010
Supplier Number:  464060010
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 ml
  • Cat. No.:
    TS464060010

Specifications

About this item

 :
 :150.97 g/mol
UN:1591
CAS Number:2199-69-1
Formula:C₆D₄Cl₂
MDL Number:MFCD00037106