Order Entry
Puerto Rico
ContactUsLinkComponent
Chloroform-[D₁] 99.8+ atom % D + 0.02-0.04 % v/v TMS for NMR spectroscopy
Chloroform-[D₁] 99.8+ atom % D + 0.02-0.04 % v/v TMS for NMR spectroscopy
Catalog # TS464020075
New Product
CAS Number:  
Chloroform-[D₁] 99.8+ atom % D + 0.02-0.04 % v/v TMS for NMR spectroscopy
Catalog # TS464020075
Supplier Number:  464020075
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    7500 ul
  • Cat. No.:
    TS464020075

Specifications

About this item

 :
 :120.38 g/mol
UN:1888
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827