Order Entry
Puerto Rico
ContactUsLinkComponent
Bromobenzene-[D₅] 99.5 atom % D for NMR spectroscopy
Bromobenzene-[D₅] 99.5 atom % D for NMR spectroscopy
Catalog # TS321230100
New Product
CAS Number:  
Bromobenzene-[D₅] 99.5 atom % D for NMR spectroscopy
Catalog # TS321230100
Supplier Number:  321230100
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 ml
  • Cat. No.:
    TS321230100

Specifications

About this item

 :
 :161.97 g/mol
UN:2514
CAS Number:4165-57-5
Formula:C₆D₅Br
MDL Number:MFCD00000056