Order Entry
Puerto Rico
ContactUsLinkComponent
Toluene-[D₈] 99.5 atom % D for NMR spectroscopy
Toluene-[D₈] 99.5 atom % D for NMR spectroscopy
Catalog # TS320790075
New Product
CAS Number:  
Toluene-[D₈] 99.5 atom % D for NMR spectroscopy
Catalog # TS320790075
Supplier Number:  320790075
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass ampoule
  • Size:
    0.75 ml
  • Cat. No.:
    TS320790075

Specifications

About this item

 :
 :100.08 g/mol
UN:1294
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638