Order Entry
Puerto Rico
ContactUsLinkComponent
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Catalog # TS214550100
New Product
CAS Number:  
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Catalog # TS214550100
Supplier Number:  214550100
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 ml
  • Cat. No.:
    TS214550100

Specifications

About this item

 :
 :86.92 g/mol
UN:1593
CAS Number:1665-00-5
Formula:CD₂Cl₂
MDL Number:MFCD00000882