

Hydrochloric acid ∼32% in water for analysis
Catalog # TS389300025
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:2500 ML
- Cat. No.:TS389300025
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Residue after ign. (sulfated):≤0.0005 %
- Titration with NaOH:≥32 %
- Bromide (Br):≤0.005 %
- Sulfite (SO3):≤0.0001 %
- Sulfate (SO4):≤0.0001 %
- Aluminium (Al):≤1 ppm
- Ammonium (NH4):≤0.0003 %
- Arsenic (As):≤0.1 ppm
- Barium (Ba):≤0.5 ppm
- Cadmium (Cd):≤0.01 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.01 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.2 ppm
- Lead (Pb):≤0.02 ppm
- Lithium (Li):≤0.02 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.01 ppm
- Nickel (Ni):≤0.02 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.5 ppm
- Strontium (Sr):≤0.02 ppm
- Zinc (Zn):≤0.05 ppm
- Heavy metals (as Pb):≤1 ppm
Specification Test Results
About this item
:
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.16 g/cm³
Storage Temperature:Ambient
ADR:8,II
:36.46
UN:1789
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324