


Hydrogen chloride ∼0.5 M in methanol, AcroSeal®
Catalog # TS428931000
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:TS428931000
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Strength:0.4 to 0.6 M
- Melting point:143 to 149 °C
- Titration Argentometric:≥94 %
Specification Test Results
About this item
:
:36.46
Density:0.8 g/cm³
Flash Pt:12 °C
Storage Temperature:Refrigerator
UN:3286
ADR:3,II
:GHS02
:GHS06
:GHS08
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324