


Hydrogen chloride 1 M in acetic acid, AcroSeal®
Catalog # TS370271000
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:TS370271000
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:1 M Exact strength on the certificate of analysis
Specification Test Results
About this item
:
:36.46 g/mol
Density:1.092 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
UN:2920
ADR:8,II
:GHS07
:GHS05
:GHS02
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324