Order Entry
Puerto Rico
ContactUsLinkComponent
Bromine 1 mol/l in acetic acid, AcroSeal®
Toxic
Corrosive
Environment
Flammable
Bromine 1 mol/l in acetic acid, AcroSeal®
Catalog # TS388401000
New Product
CAS Number:  
Toxic
Corrosive
Environment
Flammable
Bromine 1 mol/l in acetic acid, AcroSeal®
Catalog # TS388401000
Supplier Number:  388401000
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    TS388401000

Specifications

Specification Test Results

  • Appearance:
    Clear red Solution
  • Strength:
    0.90 to 1.10 M

Specification Test Results

About this item

 :
 :159.81 g/mol
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:231-778-1
UN:2922
ADR:8,I
 :GHS06
 :GHS05
 :GHS09
 :GHS02
CAS Number:7726-95-6
Formula:Br₂
MDL Number:MFCD00010896