


Tribromoboranemethyl sulfide 1 M in methylene chloride, AcroSeal™
Catalog # TS428321000
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:TS428321000
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Strength:0.95 to 1.10 M
- Melting point:185 to 189 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:312.652 g/mol
Density:1.456 g/cm³
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS06
:GHS08
:GHS05
CAS Number:29957-59-3
Formula:C₂H₆BBr₃S