Order Entry
Puerto Rico
ContactUsLinkComponent
Benzyl chloroformate 50% (w/w) 3 M in toluene, AcroSeal®
Irritant
Health
Corrosive
Environment
Flammable
Benzyl chloroformate 50% (w/w) 3 M in toluene, AcroSeal®
Catalog # TS428901000
New Product
CAS Number:  
Irritant
Health
Corrosive
Environment
Flammable
Benzyl chloroformate 50% (w/w) 3 M in toluene, AcroSeal®
Catalog # TS428901000
Supplier Number:  428901000
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    TS428901000

Specifications

Specification Test Results

  • Appearance:
    Clear light yellow to light pink Solution
  • GC:
    45 to 53 % w/w

Specification Test Results

About this item

 :
 :170.6 g/mol
Density:1.01 g/cm³
Flash Pt:7 °C
Storage Temperature:Refrigerator
UN:2920
ADR:8,I
 :GHS07
 :GHS08
 :GHS05
 :GHS09
 :GHS02
CAS Number:501-53-1
Formula:C₈H₇ClO₂
MDL Number:MFCD00000640