1-Hexadecyl-3-methylimidazolium chloride monohydrate 98%
Catalog # TS356650100
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:10 g
- Environmentally Preferable:
- Cat. No.:TS356650100
Specifications
Specification Test Results
- Appearance:Light yellow Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Water:3 to 5 % (K.F.)
Specification Test Results
About this item
Synonyms: 1-Hexadecyl-3-methyl-1H-imidazol-3-ium chloride hydrate
:
:361.01 g/mol
:GHS07
CAS Number:404001-62-3
Formula:C₂₀H₃₉ClN₂·1H₂O
MDL Number:MFCD07773044