Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Isobutanol for HPLC

Glass bottle, 1 L
Catalog # TS412640010
Irritant
Corrosive
Flammable
 :  412640010
Isobutanol for HPLC
Catalog # TS412640010
CAS Number:  78-83-1
Isobutanol for HPLC

Glass bottle, 1 L

CAS Number:  78-83-1
Price
$176.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
  • CAS Number:78-83-1
  • Formula:(CH₃)₂CHCH₂OH
  • MDL Number:MFCD00004740
  • Molecular Weight:74.12 g/mol
  • Boiling Pt:108 °C (760 mmHg)
  • Melting Pt:–108 °C
  • Density:0.8 g/cm³
  • Flash Pt:28 °C
  • Storage Temperature:Ambient
  • UN:1212
  • ADR:3,III
Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.45 %
  • UV:
    at 219 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.