Order Entry
United States
ContactUsLinkComponent
Dimethyl sulphoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Catalog # TS325370050
New Product
CAS Number:  
Dimethyl sulphoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Catalog # TS325370050
Supplier Number:  325370050
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass ampoule
  • Size:
    5 ml
  • Environmentally Preferable:
  • Cat. No.:
    TS325370050

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.85 %

Specification Test Results

About this item

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6
 :
 :84.09 g/mol
Boiling Pt:189
Melting Pt:20.2
Density:1.19 g/cm³
Flash Pt:88 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-617-0
CAS Number:2206-27-1
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090