Order Entry
Puerto Rico
ContactUsLinkComponent
Glutaraldehyde 25% in water for electron microscopy, purified
Corrosive
Irritant
Health
Environment
Glutaraldehyde 25% in water for electron microscopy, purified
New Product
Certificates
Corrosive
Irritant
Health
Environment
Glutaraldehyde 25% in water for electron microscopy, purified
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Clear to slightly hazy solution
  • Assay:
    25 to 27 % w/w
  • Color scale:
    ≤100 APHA
  • Specific gravity:
    (20/20°C) 1.000 to 1.133
  • pH:
    3.1 to 4.5 (at 25°C)

Specification Test Results