Order Entry
Puerto Rico
ContactUsLinkComponent
Pyridine for HPLC
Irritant
Flammable
Pyridine for HPLC
Catalog # TS396800010
New Product
CAS Number:  
Irritant
Flammable
Pyridine for HPLC
Catalog # TS396800010
Supplier Number:  396800010
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    TS396800010

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.8 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Color scale:
    ≤20 APHA
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

About this item

 :
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–42 °C
Density:0.978 g/cm³
Flash Pt:17 °C
Storage Temperature:Ambient
UN:1282
ADR:3,II
 :79.1 g/mol
 :GHS07
 :GHS02
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732