



Antimony(V) chloride 1M in methylene chloride, AcroSeal®
Catalog # TS434521000
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:TS434521000
Specifications
Specification Test Results
- Appearance:Light yellow to dark yellow Solution
- Titration Argentometric:0.95 to 1.10 M
- Refractive index:1.5310 to 1.5350 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:299.02 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:3 °C
Density:1.442 g/cm³
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:7647-18-9
Formula:Cl₅Sb
MDL Number:MFCD00011213