Order Entry
Puerto Rico
ContactUsLinkComponent
Toluene 99.85% for pesticide residue analysis
Toluene 99.85% for pesticide residue analysis
Catalog # TS332070010
New Product
CAS Number:  
Toluene 99.85% for pesticide residue analysis
Catalog # TS332070010
Supplier Number:  332070010
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    TS332070010

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (K.F.)
  • GC:
    ≥99.80 %
  • GC/ECD (as lindane):
    ≤5 ng/l
  • Residue after evaporation:
    ≤0.0005 %

Specification Test Results

About this item

 :
 :92.14 g/mol
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.866 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
 :GHS07
 :GHS08
 :GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512