Order Entry
Puerto Rico
ContactUsLinkComponent
2-Methyl-2-butene 2 M in THF, AcroSeal™
Irritant
Health
Flammable
2-Methyl-2-butene 2 M in THF, AcroSeal™
Catalog # TS426391000
New Product
CAS Number:  
Irritant
Health
Flammable
2-Methyl-2-butene 2 M in THF, AcroSeal™
Catalog # TS426391000
Supplier Number:  426391000
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    TS426391000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Strength:
    1.90 to 2.20 M
  • Melting point:
    107 to 110 °C
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :70.13 g/mol
Boiling Pt:39 °C (1013 hPa)
Melting Pt:–124 °C
Density:0.835 g/cm³
Flash Pt:–46 °C
UN:1993
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:513-35-9
 :26760-64-5
Formula:C₅H₁₀
MDL Number:MFCD00009276