Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
2,2,4-Trimethylpentane 99.5% for analysis

2,2,4-Trimethylpentane 99.5% for analysis

Irritant
Health
Environment
Flammable
2,2,4-Trimethylpentane 99.5% for analysis
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
  • CAS Number:540-84-1
  • Formula:(CH₃)₂CHCH₂C(CH₃)₃
  • MDL Number:MFCD00008943
  • Molecular Weight:114.23 g/mol
  • Boiling Pt:98…99 °C (760 mmHg)
  • Melting Pt:–107 °C
  • Density:0.69 g/cm³
  • Flash Pt:–12 °C
  • Storage Temperature:Ambient
  • EINECS:208-759-1
  • UN:1262
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 % (Coulometric)
  • GC:
    ≥99.4 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Aluminium (Al):
    ≤0.5 ppm
  • Cadmium (Cd):
    ≤0.1 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Nickel (Ni):
    ≤0.1 ppm
  • Zinc (Zn):
    ≤0.1 ppm
  • H2S+foreign org. S cmpds:
    ≤50 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    2500 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $176.00
    $176.00
    Each(1/Itm/Bx)
       $176.00

    Supplier Number: 265440025
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $95.00
    $95.00
    Each(1/Itm/Bx)
       $95.00

    Supplier Number: 265440010
  • Metal drum
    25 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $1,330.00
    $1,330.00
    Each(1/Itm/Bx)
       $1,330.00

    Supplier Number: 265440250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.