Order Entry
Puerto Rico
ContactUsLinkComponent
4-Methyl-2-pentanone 99.5% for HPLC
Irritant
Flammable
4-Methyl-2-pentanone 99.5% for HPLC
Catalog # TS433570010
New Product
CAS Number:  
Irritant
Flammable
4-Methyl-2-pentanone 99.5% for HPLC
Catalog # TS433570010
Supplier Number:  433570010
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    TS433570010

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.4 %
  • UV:
    at 400 nm A: ≤0.01
  • Color scale:
    ≤15 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Acidity/alkalinity:
    ≤0.0005 meq/g

Specification Test Results

About this item

 :
 :100.16 g/mol
Boiling Pt:117.4 °C (760 mmHg)
Melting Pt:–84 °C
Density:0.8 g/cm³
Flash Pt:14 °C
UN:1245
ADR:3,II
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938