Order Entry
Puerto Rico
ContactUsLinkComponent
n-Nonane, anhydrous 99%, AcroSeal®
Flammable
Irritant
Health
Environment
n-Nonane, anhydrous 99%, AcroSeal®
Catalog # TS435631000
New Product
CAS Number:  
Flammable
Irritant
Health
Environment
n-Nonane, anhydrous 99%, AcroSeal®
Catalog # TS435631000
Supplier Number:  435631000
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    TS435631000

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4040 to 1.4060 (20°C, 589 nm)
  • Water:
    ≤0.005 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specification Test Results

About this item

 :
Boiling Pt:151 °C (760 mmHg)
Melting Pt:–53 °C
Density:0.718 g/cm³
Flash Pt:31 °C
Storage Temperature:Ambient
UN:1920
ADR:3,III
 :128.26 g/mol
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:111-84-2
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574