Order Entry
Puerto Rico
ContactUsLinkComponent
Diethyl ether, extra dry 99.5% stabilized, AcroSeal®
Irritant
Flammable
Diethyl ether, extra dry 99.5% stabilized, AcroSeal®
Catalog # TS448421000
New Product
CAS Number:  
Irritant
Flammable
Diethyl ether, extra dry 99.5% stabilized, AcroSeal®
Catalog # TS448421000
Supplier Number:  448421000
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    TS448421000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.4 %
  • Stabilizer:
    1.5 to 10 ppm BHT
  • Residue after evaporation:
    ≤0.001 %
  • Acidity:
    ≤0.0002 meq/g
  • Filt.over 0.2 micron PFTE:
    Filtered
  • Peroxides:
    ≤0.002 % (as H2O2)

Specification Test Results

About this item

 :
 :74.12 g/mol
Boiling Pt:34.6 °C (760 mmHg)
Melting Pt:–116 °C
Density:0.714 g/cm³
Flash Pt:–45 °C
UN:1155
ADR:3,I
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646