Order Entry
Puerto Rico
ContactUsLinkComponent
Oberon 12 Cal TCG™ Arc Flash Face Shield and Hard Cap
Oberon 12 Cal TCG™ Arc Flash Face Shield and Hard Cap
Catalog # 77575-524
Supplier:  Sellstrom
New Product
undefined
Oberon 12 Cal TCG™ Arc Flash Face Shield and Hard Cap
Catalog # 77575-524
Supplier:  Sellstrom
Supplier Number:  21AGR12AF-C+500
New Product

Specifications

  • Color:
    Clear grey
  • Dimensions:
    17×8×8"
  • Material:
    Polycarbonate
  • Cat. No.:
    77575-524
  • Weight:
    2.83 lbs.

Specifications

About this item

Shield made from a durable polycarbonate and has anti-fog and scratch-resistant coatings.

Face shield comes equipped with an Oberon or Canadian hard cap.

Certifications: The Oberon 12 Cal Arc Flash Face Shield meets ANSI /ISEA 125 Level 2 Conformity and Arc Flash PPE Category 2 Standards with an arc rating of 17 cal/cm2. Please refer to NFPA 70E or CSA Z462 Standards for specific selection requirements.