ContactUsLinkComponent
NANOCOLOR® Photometric Tube Tests, Sludge Activity
NANOCOLOR® Photometric Tube Tests, Sludge Activity
Catalog # 77578-596
Supplier:  Macherey-Nagel
New Product
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Sludge Activity
Catalog # 77578-596
Supplier:  Macherey-Nagel
Supplier Number:  985890
New Product
CAS Number:  
Specifications
  • Chemical method:
    2,3,5-Triphenyltetrazoliumchloride (TTC)
  • Description:
    TTC/sludge activity 150
  • Cat. No.:
    77578-596
  • Range:
    5 - 150 µg TPF
  • No. of tests:
    20
Specifications
About this item
A pre-dosed NANOCOLOR® tube test for photometric determination of sludge activity in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.