NANOCOLOR® Photometric Tube Tests, AOX
Catalog # 77578-430
Supplier: Macherey-Nagel
New Product
CAS Number:
Specifications
- Chemical method:Mercury(II) -thiocyanate/Iron(III) -nitrate
- Description:AOX 3
- Cat. No.:77578-430
- Range:0.1 - 3.0 mg/L AOX
0.01 - 0.30 mg/L AOX - No. of tests:20 tests
Specifications
About this item
A pre-dosed NANOCOLOR® tube test for photometric determination of AOX in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (Ext.Ø) tubes