Orders LinkContactUsLinkComponent
Bottle Liquid Sampler
Bottle Liquid Sampler
Bottle Liquid Sampler
Certificates
Bottle Liquid Sampler
2 Options Available Below
Product Details & Documents
Incorporates a spring loaded mechanism, which means that liquid only flows in when the handle is squeezed allowing samples to be taken from set locations. Sampling Systems™ bottle liquid sampler takes samples straight into the bottle.
  • 316L Stainless Steel sampler body
  • Glass bottle
  • Single point
  • Sampling for low to medium viscosity liquids
  • Ideal for low volume (unit dose) sampling
The sampler is available with a cover that stops the bottle getting wet.
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 77576-434
    Bottle liquid sampler with 500 ml glass bottle, bottle cover
    Bottle liquid sampler with 500 ml glass bottle, bottle cover
    500 ml
    1000 mm
    Discontinued
    Supplier Number: 5031C-1000
  • 77576-438
    Bottle liquid sampler with 500 ml glass bottle
    Bottle liquid sampler with 500 ml glass bottle
    500 ml
    1000 mm
    Log in for availability
    Log in for availability
    Each
    Each - $2,100.00
    $2,100.00
    Each
       $2,100.00
    Supplier Number: 5031S-1000
  • 77576-436
    Bottle liquid sampler with 500 ml glass bottle, bottle cover
    Bottle liquid sampler with 500 ml glass bottle, bottle cover
    500 ml
    1500 mm
    Discontinued
    Supplier Number: 5031C-1500
  • 77576-440
    Bottle liquid sampler with 500 ml glass bottle
    Bottle liquid sampler with 500 ml glass bottle
    500 ml
    1500 mm
    Log in for availability
    Log in for availability
    Each
    Each - $2,310.00
    $2,310.00
    Each
       $2,310.00
    Supplier Number: 5031S-1500